siRNA Details
| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033079.1 | NP_001028251.1 | 12 | 4427 | ||
| GenBank mRNA | L06434.2 | - | 12 | 4427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a17| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033079.1 | NP_001028251.1 | 12 | 4427 | ||
| GenBank mRNA | L06434.2 | - | 12 | 4427 | ||