siRNA Details
| Gene Symbol: Surf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033868.1 | NP_001029040.1 | 6 | 990 | ||
| GenBank mRNA | CO558944.1 | - | 1 | 311 | ||
| Gene Symbol: LOC102553670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233722.2 | XP_006233784.1 | 5 | 2842 | ||

Surf4
Genomic Map