siRNA Details
| Gene Symbol: Ppapdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034854.1 | NP_001030026.1 | 1 | 2160 | ||
| GenBank mRNA | BC081970.1 | - | 1 | 2160 | 7125531 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppapdc2| Gene Symbol: Ppapdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034854.1 | NP_001030026.1 | 1 | 2160 | ||
| GenBank mRNA | BC081970.1 | - | 1 | 2160 | 7125531 | |