siRNA Details
| Gene Symbol: Gpr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034855.1 | NP_001030027.1 | 2637 | |||
| GenBank mRNA | BC105872.1 | - | 2637 | 7133996 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr153| Gene Symbol: Gpr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034855.1 | NP_001030027.1 | 2637 | |||
| GenBank mRNA | BC105872.1 | - | 2637 | 7133996 | ||