siRNA Details
| Gene Symbol: Cdca7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034953.1 | NP_001030125.1 | 10 | 1750 | ||
| GenBank mRNA | BC098734.1 | - | 10 | 1750 | 7385105 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdca7l| Gene Symbol: Cdca7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034953.1 | NP_001030125.1 | 10 | 1750 | ||
| GenBank mRNA | BC098734.1 | - | 10 | 1750 | 7385105 | |