siRNA Details
| Gene Symbol: Tmem69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035001.2 | NP_001030173.1 | 2, 3 | 177 | ||
| GenBank mRNA | BC083906.1 | - | 382 | 7133962 | ||
| BC161923.1 | - | 2, 3 | 177 | 8370867 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem69| Gene Symbol: Tmem69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035001.2 | NP_001030173.1 | 2, 3 | 177 | ||
| GenBank mRNA | BC083906.1 | - | 382 | 7133962 | ||
| BC161923.1 | - | 2, 3 | 177 | 8370867 | ||