siRNA Details
| Gene Symbol: Hibadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022243.1 | NP_071579.1 | 7 | 737 | ||
| GenBank mRNA | BC127442.1 | - | 7 | 723 | 7308860 | |
| J04628.1 | - | 7 | 738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hibadh| Gene Symbol: Hibadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022243.1 | NP_071579.1 | 7 | 737 | ||
| GenBank mRNA | BC127442.1 | - | 7 | 723 | 7308860 | |
| J04628.1 | - | 7 | 738 | |||