siRNA Details
| Gene Symbol: Nup62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023098.2 | NP_075586.1 | 2 | 801 | ||
| XM_006229150.2 | XP_006229212.1 | 2 | 1000 | |||
| XM_006229151.2 | XP_006229213.1 | 2 | 1706 | |||
| GenBank mRNA | BC128709.1 | - | 1 | 705 | 8363131 | |
| X52583.1 | - | 1 | 1381 | |||

Nup62
Genomic Map