siRNA Details
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031025.1 | NP_112287.1 | 14 | 1976 | ||
| GenBank mRNA | BC107440.1 | - | 14 | 1976 | 7115402 | |
| D10655.1 | - | 13 | 1693 | |||
| FQ234755.1 | - | 2 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlat| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031025.1 | NP_112287.1 | 14 | 1976 | ||
| GenBank mRNA | BC107440.1 | - | 14 | 1976 | 7115402 | |
| D10655.1 | - | 13 | 1693 | |||
| FQ234755.1 | - | 2 | 280 | |||