siRNA Details
| Gene Symbol: Pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031086.2 | NP_112348.2 | 8, 9 | 931 | ||
| XM_006240719.2 | XP_006240781.1 | 6, 7 | 1217 | |||
| XM_008765045.1 | XP_008763267.1 | 6, 7 | 663 | |||
| GenBank mRNA | CB580854.1 | - | 3, 4 | 288 | ||

Pros1
Genomic Map