siRNA Details
| Gene Symbol: Pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031086.2 | NP_112348.2 | 15 | 1980 | ||
| XM_006240719.2 | XP_006240781.1 | 13 | 2266 | |||
| XM_008765045.1 | XP_008763267.1 | 13 | 1712 | |||
| GenBank mRNA | CK366505.1 | - | 3 | 405 | ||

Pros1
Genomic Map