siRNA Details
| Gene Symbol: SLC17A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020309.3 | NP_064705.1 | 3 | 580 | ||
| GenBank mRNA | AB032436.1 | - | 3 | 429 | ||
| AK057879.1 | - | 526 | ||||
| AK095659.2 | - | 2 | 1859 | |||
| AK294226.1 | - | 3 | 386 | |||
| AK294405.1 | - | 3 | 429 | |||
| BC059379.1 | - | 3 | 555 | 5262415 | ||
| BG701511.1 | - | 3 | 556 | |||

SLC17A7
Genomic Map