siRNA Details
| Gene Symbol: LOC103693620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769033.1 | XP_008767255.1 | 1 | 285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103693620| Gene Symbol: LOC103693620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769033.1 | XP_008767255.1 | 1 | 285 | ||