siRNA Details
| Gene Symbol: Ube2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037292.1 | NP_001032369.1 | 6 | 547 | ||
| GenBank mRNA | U13176.1 | - | 6 | 547 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2d2| Gene Symbol: Ube2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037292.1 | NP_001032369.1 | 6 | 547 | ||
| GenBank mRNA | U13176.1 | - | 6 | 547 | ||