siRNA Details
| Gene Symbol: Popdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037369.1 | NP_001032446.1 | 3 | 683 | ||
| XM_006256599.2 | XP_006256661.1 | 2 | 923 | |||
| GenBank mRNA | AY787642.1 | - | 1 | 650 | ||
| BC105761.1 | - | 3 | 683 | 5623862 | ||
| FQ218039.1 | - | 3 | 585 | |||

Popdc3
Genomic Map