siRNA Details
| Gene Symbol: Lrrc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037336.1 | NP_001032413.1 | 1 | 350 | ||
| GenBank mRNA | DQ119102.1 | - | 1 | 350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc4| Gene Symbol: Lrrc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037336.1 | NP_001032413.1 | 1 | 350 | ||
| GenBank mRNA | DQ119102.1 | - | 1 | 350 | ||