siRNA Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037664.1 | NP_001032753.1 | 9 | 1089 | ||
| GenBank mRNA | AY623026.1 | - | 1663 | |||
| BC088267.1 | - | 9 | 1089 | 7379090 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtnbp1| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037664.1 | NP_001032753.1 | 9 | 1089 | ||
| GenBank mRNA | AY623026.1 | - | 1663 | |||
| BC088267.1 | - | 9 | 1089 | 7379090 | ||