siRNA Details
| Gene Symbol: Defb51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037547.1 | NP_001032636.1 | 3 | 370 | ||
| GenBank mRNA | AY621373.1 | - | 3 | 370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb51| Gene Symbol: Defb51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037547.1 | NP_001032636.1 | 3 | 370 | ||
| GenBank mRNA | AY621373.1 | - | 3 | 370 | ||