siRNA Details
| Gene Symbol: Ggta1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037583.1 | NP_001032672.1 | 5 | 400 | ||
| XM_006234046.2 | XP_006234108.1 | 6 | 434 | |||
| GenBank mRNA | AY524048.1 | - | 5 | 400 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggta1l1| Gene Symbol: Ggta1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037583.1 | NP_001032672.1 | 5 | 400 | ||
| XM_006234046.2 | XP_006234108.1 | 6 | 434 | |||
| GenBank mRNA | AY524048.1 | - | 5 | 400 | ||