siRNA Details
| Gene Symbol: Rbm12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037657.1 | NP_001032746.1 | 1381 | |||
| XM_008762382.1 | XP_008760604.1 | 3 | 1441 | |||
| GenBank mRNA | AY226099.1 | - | 1381 | |||
| Gene Symbol: LOC100910990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235453.2 | XP_006235515.1 | 3 | 1409 | ||
| XM_008762412.1 | XP_008760634.1 | 3 | 1441 | |||
| XM_008762414.1 | XP_008760636.1 | 2 | 3404 | |||

Rbm12
Genomic Map