siRNA Details
| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037660.1 | NP_001032749.1 | 5 | 419 | ||
| GenBank mRNA | AJ628424.1 | - | 5 | 419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csf2ra| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037660.1 | NP_001032749.1 | 5 | 419 | ||
| GenBank mRNA | AJ628424.1 | - | 5 | 419 | ||