siRNA Details
| Gene Symbol: Spag11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037850.1 | NP_001032939.1 | 2 | 95 | ||
| NM_001037852.1 | NP_001032941.1 | 2 | 155 | |||
| GenBank mRNA | DQ012092.1 | - | 2 | 95 | ||
| DQ012093.1 | - | 2 | 155 | |||

Spag11b
Genomic Map