siRNA Details
| Gene Symbol: Defb27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037519.1 | NP_001032608.1 | 2 | 251 | ||
| GenBank mRNA | AY600149.1 | - | 2 | 258 | ||
| AY621358.1 | - | 2 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb27| Gene Symbol: Defb27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037519.1 | NP_001032608.1 | 2 | 251 | ||
| GenBank mRNA | AY600149.1 | - | 2 | 258 | ||
| AY621358.1 | - | 2 | 251 | |||