siRNA Details
| Gene Symbol: Cep112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029586.2 | NP_083862.1 | 3 | 431 | ||
| NM_029606.3 | NP_083882.3 | 26 | 2978 | |||
| NM_145688.2 | NP_663721.2 | 5 | 653 | |||
| GenBank mRNA | AK005611.1 | - | 3 | 432 | ||
| AK006504.1 | - | 5 | 639 | |||
| BC005723.1 | - | 5 | 465 | |||
| BC048677.1 | - | 5 | 625 | |||
| BC086808.1 | - | 20 | 2218 | 30632964 | ||
| CA464637.1 | - | 5 | 655 | |||

Cep112
Genomic Map