siRNA Details
| Gene Symbol: Hdac9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024124.3 | NP_077038.2 | 12 | 2965 | ||
| GenBank mRNA | AK144242.1 | - | 3 | 1438 | ||
| BC098187.1 | - | 12 | 2951 | 5363624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdac9| Gene Symbol: Hdac9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024124.3 | NP_077038.2 | 12 | 2965 | ||
| GenBank mRNA | AK144242.1 | - | 3 | 1438 | ||
| BC098187.1 | - | 12 | 2951 | 5363624 | ||