siRNA Details
| Gene Symbol: Mettl7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027334.3 | NP_081610.2 | 2 | 1593 | ||
| GenBank mRNA | AB253943.1 | - | 2 | 1561 | ||
| AJ536232.1 | - | 2 | 1565 | |||
| AK076169.1 | - | 2 | 1595 | |||
| BC033446.1 | - | 2 | 1570 | |||
| BC047149.1 | - | 2 | 1578 | |||
| BC055034.1 | - | 2 | 1584 | 6306873 | ||
| BC060144.1 | - | 2 | 1573 | |||

Mettl7a1
Genomic Map