siRNA Details
| Gene Symbol: Vmn1r30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134177.1 | NP_598938.1 | 1 | 875 | ||
| GenBank mRNA | BC138075.1 | - | 1 | 924 | 8861094 | |
| BC138078.1 | - | 1 | 924 | 8861097 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r30| Gene Symbol: Vmn1r30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134177.1 | NP_598938.1 | 1 | 875 | ||
| GenBank mRNA | BC138075.1 | - | 1 | 924 | 8861094 | |
| BC138078.1 | - | 1 | 924 | 8861097 | ||