siRNA Details
| Gene Symbol: Olfr767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146318.2 | NP_666430.2 | 1 | 223 | ||
| GenBank mRNA | BC137767.1 | - | 1 | 319 | 8860786 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr767| Gene Symbol: Olfr767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146318.2 | NP_666430.2 | 1 | 223 | ||
| GenBank mRNA | BC137767.1 | - | 1 | 319 | 8860786 | |