siRNA Details
| Gene Symbol: Olfr531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146953.1 | NP_667164.1 | 1 | 65 | ||
| GenBank mRNA | BC104052.1 | - | 1 | 65 | 40040241 | |
| BC104053.1 | - | 1 | 65 | 40040242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr531| Gene Symbol: Olfr531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146953.1 | NP_667164.1 | 1 | 65 | ||
| GenBank mRNA | BC104052.1 | - | 1 | 65 | 40040241 | |
| BC104053.1 | - | 1 | 65 | 40040242 | ||