siRNA Details
| Gene Symbol: Cyp4a12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172306.2 | NP_758510.2 | 12 | 2344 | ||
| GenBank mRNA | AK098123.1 | - | 12 | 2325 | ||
| BC060945.1 | - | 12 | 2325 | 30304458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4a12b| Gene Symbol: Cyp4a12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172306.2 | NP_758510.2 | 12 | 2344 | ||
| GenBank mRNA | AK098123.1 | - | 12 | 2325 | ||
| BC060945.1 | - | 12 | 2325 | 30304458 | ||