siRNA Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172909.4 | NP_766497.3 | 8 | 2541 | ||
| GenBank mRNA | AF539744.1 | - | 8 | 2297 | ||
| AK046675.1 | - | 7 | 2665 | |||
| AK088010.1 | - | 8 | 2528 | |||
| BC145639.1 | - | 7 | 2390 | |||
| BC145640.1 | - | 7 | 2390 | |||

Cd163l1
Genomic Map