siRNA Details
| Gene Symbol: Xlr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011727.2 | NP_035857.2 | 5, 6 | 393 | ||
| GenBank mRNA | L22977.1 | - | 378 | |||
| U02600.1 | - | 5, 6 | 393 | |||
| Gene Symbol: Xlr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081643.1 | NP_001075112.1 | 5, 6 | 424 | ||
| GenBank mRNA | BC038862.1 | - | 5, 6 | 387 | ||
| BC064787.1 | - | 5, 6 | 381 | 30046437 | ||

Xlr3c
Genomic Map