siRNA Details
| Gene Symbol: Xlr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081643.1 | NP_001075112.1 | 2 | 118 | ||
| GenBank mRNA | AK146398.1 | - | 2 | 87 | ||
| BC038862.1 | - | 2 | 81 | |||
| BC057003.1 | - | 2 | 90 | |||
| BC064787.1 | - | 2 | 75 | 30046437 | ||
| CJ063846.1 | - | 2 | 119 | |||
| Gene Symbol: Xlr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011727.2 | NP_035857.2 | 2 | 87 | ||
| GenBank mRNA | L22977.1 | - | 72 | |||
| U02600.1 | - | 2 | 87 | |||

Xlr3b
Genomic Map