siRNA Details
| Gene Symbol: Il11ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U69491.1 | - | 1949 | |||
| X98519.1 | - | 1961 | ||||
| Gene Symbol: Il11ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC004619.1 | - | 1681 | 3585497 | ||
| BC069984.1 | - | 1744 | 6508165 | |||

Il11ra2
Genomic Map