siRNA Details
| Gene Symbol: Mzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290452.1 | NP_001277381.1 | 2 | 575 | ||
| NM_001290453.1 | NP_001277382.1 | 2 | 575 | |||
| NM_145819.2 | NP_665818.2 | 2 | 575 | |||
| GenBank mRNA | AB007407.1 | - | 1 | 794 | ||
| AB088478.1 | - | 2 | 476 | |||
| AK030410.1 | - | 2 | 576 | |||
| AK145014.1 | - | 2 | 456 | |||
| AK158837.1 | - | 1 | 1451 | |||
| BC060649.1 | - | 2 | 535 | |||
| BC129904.1 | - | 2 | 325 | 40111654 | ||
| BC129905.1 | - | 2 | 325 | 40111655 | ||

Mzf1
Genomic Map