siRNA Details
| Gene Symbol: Farp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134082.3 | NP_598843.3 | 21 | 2773 | ||
| GenBank mRNA | AK034903.1 | - | 2 | 76 | ||
| AK199091.1 | - | 1 | 36 | |||
| BB840008.1 | - | 4 | 374 | |||
| BC027077.1 | - | 4 | 316 | |||
| BC030329.1 | - | 14 | 1761 | |||
| BC043327.1 | - | 3 | 177 | |||
| BC141229.1 | - | 20 | 2334 | |||

Farp1
Genomic Map