siRNA Details
| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081012.1 | NP_001074481.1 | 26 | 4337 | ||
| GenBank mRNA | AK039282.1 | - | 14 | 3033 | ||
| BC039810.1 | - | 8 | 1176 | |||
| BC172165.1 | - | 25 | 4128 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc171| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081012.1 | NP_001074481.1 | 26 | 4337 | ||
| GenBank mRNA | AK039282.1 | - | 14 | 3033 | ||
| BC039810.1 | - | 8 | 1176 | |||
| BC172165.1 | - | 25 | 4128 | |||