siRNA Details
| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037665.1 | - | 1, 2 | 155 | ||
| AK039282.1 | - | 8, 9 | 2104 | |||
| BC039810.1 | - | 2, 3 | 247 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc171| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037665.1 | - | 1, 2 | 155 | ||
| AK039282.1 | - | 8, 9 | 2104 | |||
| BC039810.1 | - | 2, 3 | 247 | |||