siRNA Details
| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040106.2 | NP_001035195.1 | 21 | 7491 | ||
| NM_177762.6 | NP_808430.2 | 20 | 7248 | |||
| GenBank mRNA | AK047445.1 | - | 1 | 1144 | ||
| AK147363.1 | - | 21 | 7492 | |||
| BC033597.1 | - | 1 | 1117 | |||
| BC043125.1 | - | 10 | 5297 | |||
| BC059930.1 | - | 1 | 702 | |||

Aak1
Genomic Map