siRNA Details
| Gene Symbol: Ugt1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013701.3 | NP_038729.1 | 1 | 448 | ||
| GenBank mRNA | AK002629.1 | - | 1 | 449 | ||
| BC138676.1 | - | 1 | 447 | 8861697 | ||
| BC145969.1 | - | 1 | 447 | 40131272 | ||
| D87866.1 | - | 1 | 422 | |||

Ugt1a2
Genomic Map