siRNA Details
| Gene Symbol: Gstm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010359.2 | NP_034489.1 | 8 | 1095 | ||
| GenBank mRNA | BC044927.1 | - | 8 | 1079 | 5051815 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstm3| Gene Symbol: Gstm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010359.2 | NP_034489.1 | 8 | 1095 | ||
| GenBank mRNA | BC044927.1 | - | 8 | 1079 | 5051815 | |