siRNA Details
| Gene Symbol: Olfr235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146686.2 | NP_666897.2 | 1 | 130 | ||
| GenBank mRNA | BC132238.1 | - | 1 | 200 | 40130515 | |
| BC132388.1 | - | 1 | 200 | 40130665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr235| Gene Symbol: Olfr235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146686.2 | NP_666897.2 | 1 | 130 | ||
| GenBank mRNA | BC132238.1 | - | 1 | 200 | 40130515 | |
| BC132388.1 | - | 1 | 200 | 40130665 | ||