siRNA Details
| Gene Symbol: Olfr1436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146687.2 | NP_666898.2 | 1 | 608 | ||
| GenBank mRNA | BC127991.1 | - | 1 | 612 | 40135681 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1436| Gene Symbol: Olfr1436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146687.2 | NP_666898.2 | 1 | 608 | ||
| GenBank mRNA | BC127991.1 | - | 1 | 612 | 40135681 | |