siRNA Details
| Gene Symbol: Olfr724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146492.2 | NP_666703.2 | 1 | 943 | ||
| GenBank mRNA | BC120628.1 | - | 1 | 943 | 40129551 | |
| BC125538.1 | - | 1 | 943 | 40130053 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr724| Gene Symbol: Olfr724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146492.2 | NP_666703.2 | 1 | 943 | ||
| GenBank mRNA | BC120628.1 | - | 1 | 943 | 40129551 | |
| BC125538.1 | - | 1 | 943 | 40130053 | ||