siRNA Details
| Gene Symbol: RAB25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020387.3 | NP_065120.2 | 2 | 446 | ||
| GenBank mRNA | AF083124.1 | - | 2 | 407 | ||
| AF274025.1 | - | 2 | 425 | |||
| BC009831.2 | - | 2 | 420 | 3926839 | ||
| BC033322.1 | - | 2 | 410 | 4775725 | ||
| BP263750.1 | - | 2 | 446 | |||

RAB25
Genomic Map