siRNA Details
| Gene Symbol: Olfr1168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146531.2 | NP_666742.2 | 1 | 623 | ||
| GenBank mRNA | BC104084.2 | - | 1 | 623 | 40042096 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1168| Gene Symbol: Olfr1168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146531.2 | NP_666742.2 | 1 | 623 | ||
| GenBank mRNA | BC104084.2 | - | 1 | 623 | 40042096 | |