siRNA Details
Gene Symbol: Olfr850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146523.1 | NP_666734.1 | 1 | 530 | ||
GenBank mRNA | BC113136.1 | - | 1 | 529 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146523.1 | NP_666734.1 | 1 | 530 | ||
GenBank mRNA | BC113136.1 | - | 1 | 529 |