siRNA Details
| Gene Symbol: Olfr530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146519.1 | NP_666730.1 | 1 | 751 | ||
| GenBank mRNA | BC138081.1 | - | 1 | 809 | 8861100 | |
| BC138082.1 | - | 1 | 809 | 8861101 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr530| Gene Symbol: Olfr530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146519.1 | NP_666730.1 | 1 | 751 | ||
| GenBank mRNA | BC138081.1 | - | 1 | 809 | 8861100 | |
| BC138082.1 | - | 1 | 809 | 8861101 | ||