siRNA Details
| Gene Symbol: Olfr165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146466.1 | NP_666677.1 | 1 | 837 | ||
| GenBank mRNA | BC120624.1 | - | 1 | 898 | 40129547 | |
| BC120626.1 | - | 1 | 898 | 40129549 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr165| Gene Symbol: Olfr165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146466.1 | NP_666677.1 | 1 | 837 | ||
| GenBank mRNA | BC120624.1 | - | 1 | 898 | 40129547 | |
| BC120626.1 | - | 1 | 898 | 40129549 | ||