siRNA Details
| Gene Symbol: Olfr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146465.2 | NP_666676.1 | 1 | 136 | ||
| GenBank mRNA | BC132008.1 | - | 1 | 136 | 40130285 | |
| BC132012.1 | - | 1 | 136 | 40130289 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr108| Gene Symbol: Olfr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146465.2 | NP_666676.1 | 1 | 136 | ||
| GenBank mRNA | BC132008.1 | - | 1 | 136 | 40130285 | |
| BC132012.1 | - | 1 | 136 | 40130289 | ||